Assay Details
Gene Symbol: NMNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075834.1 | NP_001069302.1 | 5 - 6 | 688 | 102 | |
GenBank mRNA | BC119834.1 | - | 5 - 6 | 688 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
