Assay Details
Gene Symbol: FGF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076007.2 | NP_001069475.1 | 3 - 4 | 362 | 74 | |
GenBank mRNA | BC120280.1 | - | 3 - 4 | 363 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
