Assay Details
Gene Symbol: FGF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076007.2 | NP_001069475.1 | 4 - 5 | 476 | 66 | |
GenBank mRNA | BC120280.1 | - | 4 - 5 | 477 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
