Assay Details
Gene Symbol: ITPKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076011.1 | NP_001069479.1 | 1 - 2 | 1150 | 115 | |
GenBank mRNA | BC119830.1 | - | 1 - 2 | 1150 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
