Assay Details
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076143.1 | NP_001069611.1 | 7 - 8 | 669 | 145 | |
GenBank mRNA | BC119989.1 | - | 7 - 8 | 669 | 145 | |
BC149417.1 | - | 7 - 8 | 752 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
