Assay Details
Gene Symbol: ZNF814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076277.2 | NP_001069745.1 | 1 - 2 | 234 | 79 | |
GenBank mRNA | BC122798.1 | - | 1 - 2 | 235 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC122798.1, NM_001076277.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
