Assay Details
Gene Symbol: ZNF814 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076277.2 | NP_001069745.1 | 3 - 4 | 519 | 78 | |
GenBank mRNA | BC122798.1 | - | 3 - 4 | 520 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
