Assay Details
Gene Symbol: C1S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076550.1 | NP_001070018.1 | 2 - 3 | 295 | 63 | |
GenBank mRNA | BC119956.1 | - | 2 - 3 | 295 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC119956.1, NM_001076550.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
