Assay Details
Gene Symbol: RWDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076824.1 | NP_001070292.1 | 3 - 4 | 425 | 97 | |
GenBank mRNA | BC123494.1 | - | 3 - 4 | 425 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
