Assay Details
Gene Symbol: CDH18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076837.1 | NP_001070305.1 | 11 - 12 | 2183 | 87 | |
GenBank mRNA | BC123714.1 | - | 11 - 12 | 2183 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
