Assay Details
Gene Symbol: CFAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076866.1 | NP_001070334.1 | 2 - 3 | 170 | 78 | |
GenBank mRNA | BC123870.1 | - | 2 - 3 | 170 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
