Assay Details
Gene Symbol: TNFRSF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076911.1 | NP_001070379.1 | 3 - 4 | 1650 | 71 | |
GenBank mRNA | BC123657.1 | - | 3 - 4 | 1650 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
