Assay Details
Gene Symbol: MIER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076913.1 | NP_001070381.1 | 11 - 12 | 1139 | 98 | |
GenBank mRNA | BC123513.1 | - | 11 - 12 | 1139 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
