Assay Details
Gene Symbol: MIER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076913.1 | NP_001070381.1 | 8 - 9 | 831 | 150 | |
GenBank mRNA | BC123513.1 | - | 8 - 9 | 831 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
