Assay Details
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077933.2 | NP_001071401.1 | 2 - 3 | 386 | 99 | |
GenBank mRNA | BC120077.1 | - | 2 - 3 | 388 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
