Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078098.1 | NP_001071566.1 | 5 - 6 | 782 | 124 | |
GenBank mRNA | BC123636.1 | - | 5 - 6 | 782 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
