Assay Details
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080367.2 | NP_001073836.1 | 1 - 2 | 320 | 117 | |
GenBank mRNA | BC120133.1 | - | 1 - 2 | 321 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
