Assay Details
Gene Symbol: AMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173990.2 | NP_776415.1 | 5 - 6 | 775 | 83 | |
GenBank mRNA | BC123795.1 | - | 5 - 6 | 775 | 83 | |
M95605.1 | - | 5 - 6 | 774 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
