Assay Details
Gene Symbol: RAB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174447.4 | NP_776872.1 | 3 - 4 | 433 | 99 | |
GenBank mRNA | BC122723.1 | - | 3 - 4 | 434 | 99 | |
M19886.1 | - | 3 - 4 | 389 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
