Assay Details
Gene Symbol: CCR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024930.3 | NP_001020101.2 | 1 - 2 | 63 | 55 | |
GenBank mRNA | BC126587.1 | - | 1 - 2 | 64 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC126587.1, NM_001024930.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
