Assay Details
Gene Symbol: PLSCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046053.2 | NP_001039518.1 | 2 - 3 | 698 | 61 | |
GenBank mRNA | BC114676.1 | - | 2 - 3 | 346 | 61 | |
BT029864.1 | - | 2 - 3 | 698 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
