Assay Details
Gene Symbol: PLSCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046053.2 | NP_001039518.1 | 6 - 7 | 1279 | 79 | |
GenBank mRNA | BC114676.1 | - | 6 - 7 | 927 | 79 | |
BT029864.1 | - | 6 - 7 | 1279 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
