Assay Details
Gene Symbol: VAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075630.2 | NP_001069098.2 | 3 - 4 | 421 | 66 | |
GenBank mRNA | BC134553.1 | - | 3 - 4 | 421 | 66 | |
BT026540.1 | - | 3 - 4 | 412 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
