Assay Details
Gene Symbol: SLC25A44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077857.1 | NP_001071325.1 | 3 - 4 | 935 | 76 | |
GenBank mRNA | BC126559.1 | - | 3 - 4 | 935 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
