Assay Details
Gene Symbol: ABO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077926.1 | NP_001071394.1 | 4 - 5 | 266 | 140 | |
GenBank mRNA | BC126634.1 | - | 4 - 5 | 266 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
