Assay Details
Gene Symbol: PMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077981.2 | NP_001071449.1 | 11 - 12 | 2571 | 92 | |
GenBank mRNA | BC126839.1 | - | 11 - 12 | 2572 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
