Assay Details
Gene Symbol: SP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078027.1 | NP_001071495.1 | 2 - 3 | 250 | 72 | |
GenBank mRNA | BC126621.1 | - | 2 - 3 | 250 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
