Assay Details
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079580.2 | NP_001073048.1 | 3 - 4 | 513 | 99 | |
GenBank mRNA | BC122771.1 | - | 3 - 4 | 514 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
