Assay Details
Gene Symbol: ELK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081612.1 | NP_001075081.1 | 3 - 4 | 1266 | 78 | |
GenBank mRNA | BC133356.1 | - | 3 - 4 | 1266 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
