Assay Details
Gene Symbol: CLIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081727.1 | NP_001075196.1 | 4 - 5 | 634 | 118 | |
GenBank mRNA | BC133567.1 | - | 4 - 5 | 634 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
