Assay Details
Gene Symbol: CLIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081727.1 | NP_001075196.1 | 5 - 6 | 830 | 101 | |
GenBank mRNA | BC133567.1 | - | 5 - 6 | 830 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
