Assay Details
Gene Symbol: IGSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082439.1 | NP_001075908.1 | 5 - 6 | 1806 | 94 | |
GenBank mRNA | BC133513.1 | - | 5 - 6 | 1806 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
