Assay Details
Gene Symbol: LYPD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082462.2 | NP_001075931.1 | 5 - 6 | 582 | 117 | |
GenBank mRNA | BC133550.1 | - | 5 - 6 | 584 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
