Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082471.2 | NP_001075940.1 | 1 - 2 | 356 | 98 | |
GenBank mRNA | BC133574.1 | - | 1 - 2 | 357 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
