Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082471.2 | NP_001075940.1 | 2 - 3 | 681 | 122 | |
GenBank mRNA | BC133574.1 | - | 2 - 3 | 682 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
