Assay Details
Gene Symbol: LYPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083419.1 | NP_001076888.1 | 1 - 2 | 113 | 95 | |
GenBank mRNA | BC123509.1 | - | 1 - 2 | 113 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
