Assay Details
Gene Symbol: RBBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083424.1 | NP_001076893.1 | 2 - 3 | 154 | 78 | |
GenBank mRNA | BC123685.1 | - | 2 - 3 | 154 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
