Assay Details
Gene Symbol: AKT1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083434.2 | NP_001076903.1 | 2 - 3 | 482 | 144 | |
GenBank mRNA | BC133618.1 | - | 2 - 3 | 483 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
