Assay Details
Gene Symbol: UBE2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083503.1 | NP_001076972.1 | 3 - 4 | 756 | 79 | |
GenBank mRNA | BC133501.1 | - | 3 - 4 | 756 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
