Assay Details
Gene Symbol: UGT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083635.1 | NP_001077104.1 | 3 - 4 | 1201 | 96 | |
GenBank mRNA | BC134489.1 | - | 3 - 4 | 1201 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
