Assay Details
Gene Symbol: ARF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083671.1 | NP_001077140.1 | 5 - 6 | 576 | 104 | |
GenBank mRNA | BC134695.1 | - | 5 - 6 | 576 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
