Assay Details
Gene Symbol: GTF2IRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083745.1 | NP_001077214.1 | 3 - 4 | 410 | 116 | |
GenBank mRNA | BC134477.1 | - | 3 - 4 | 410 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
