Assay Details
Gene Symbol: PTOV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083780.1 | NP_001077249.1 | 8 - 9 | 1029 | 84 | |
GenBank mRNA | BC134519.1 | - | 8 - 9 | 1029 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
