Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098868.1 | NP_001092338.1 | 1 - 2 | 184 | 72 | |
GenBank mRNA | BC146025.1 | - | 1 - 2 | 184 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC146025.1, NM_001098868.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
