Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098868.1 | NP_001092338.1 | 3 - 4 | 572 | 60 | |
GenBank mRNA | BC146025.1 | - | 3 - 4 | 572 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
