Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098868.1 | NP_001092338.1 | 4 - 5 | 735 | 68 | |
GenBank mRNA | BC146025.1 | - | 4 - 5 | 735 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
