Assay Details
Gene Symbol: TNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173966.3 | NP_776391.2 | 1 - 2 | 376 | 84 | |
GenBank mRNA | AF348421.1 | - | 1 - 2 | 183 | 84 | |
BC134755.1 | - | 1 - 2 | 377 | 84 | ||
JN635499.1 | - | 1 - 2 | 214 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
