Assay Details
Gene Symbol: DCPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174695.3 | NP_777120.2 | 4 - 5 | 539 | 93 | |
GenBank mRNA | AF398237.1 | - | 3 - 4 | 547 | 93 | |
BC140689.1 | - | 4 - 5 | 539 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
