Assay Details
Gene Symbol: DOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024543.2 | NP_001019714.2 | 4 - 5 | 663 | 57 | |
GenBank mRNA | BT020685.1 | - | 4 - 5 | 663 | 57 | |
BT021556.1 | - | 4 - 5 | 645 | 57 | ||
BT029827.1 | - | 4 - 5 | 652 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
