Assay Details
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035087.2 | NP_001030259.2 | 1 - 2 | 112 | 132 | |
GenBank mRNA | BC102368.1 | - | 1 - 2 | 112 | 132 | |
DT819906.1 | - | 1 - 2 | 112 | 132 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102368.1, NM_001035087.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
